KEY181739818 Key Organics / BIONET

2-{1H-pyrrolo[2,3-b]pyridin-1-yl}acetic acid

  • MDL number MFCD09834895
  • Cas Number : 1048913-13-8
  • Molecular Weight : 176.175

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 169 - 170
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96850

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/FC-0734_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart