KEY181739821 Key Organics / BIONET

2,5-dichloropyridine-4-carbaldehyde

  • MDL number MFCD06410679
  • Cas Number : 102645-33-0
  • Molecular Weight : 176

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 73 - 74
Iupac Name 2,5-dichloroisonicotinaldehyde
Inchi Code 1S/C6H3Cl2NO/c7-5-2-9-6(8)1-4(5)3-10/h1-3H
Inchi Key UIPSRNHDSBVXHY-UHFFFAOYSA-N

Description

Melting Point 73 - 74
Purity 97%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96830

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/FC-0601_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart