KEY181739824 Key Organics / BIONET

3-chloro-5-nitropyridin-2-ol

  • MDL number MFCD06656618
  • Cas Number : 22353-38-4
  • Molecular Weight : 174.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 187 - 189
Iupac Name 3-chloro-5-nitro-2(1H)-pyridinone
Inchi Code 1S/C5H3ClN2O3/c6-4-1-3(8(10)11)2-7-5(4)9/h1-2H,(H,7,9)
Inchi Key GTOXJMNILNYXLE-UHFFFAOYSA-N

Description

Melting Point 187 - 189
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96833

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/FC-0605_SDS.pdf
hcodes H302,H312,H314,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart