KEY181739826 Key Organics / BIONET

2-hydroxy-5-iodopyridine-3-carboxylic acid

  • MDL number MFCD11100199
  • Cas Number : 390360-97-1
  • Molecular Weight : 265.006

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 264 - 266
Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94479

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AC-0201_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart