KEY181739829 Key Organics / BIONET

2-amino-5-bromopyrimidin-4-ol

  • MDL number MFCD09999234
  • Cas Number : 61937-71-1
  • Molecular Weight : 190

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 284 - 285
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96841

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/FC-0700_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart