KEY181740379 Key Organics / BIONET

5-bromo-6-hydroxypyridine-3-carbonitrile

  • MDL number MFCD11100229
  • Cas Number : 19840-44-9
  • Molecular Weight : 199.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 259 - 261
Iupac Name 5-bromo-6-hydroxynicotinonitrile
Inchi Code 1S/C6H3BrN2O/c7-5-1-4(2-8)3-9-6(5)10/h1,3H,(H,9,10)
Inchi Key NCIBXYJEJGGUFF-UHFFFAOYSA-N

Description

Melting Point 259 - 261
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97317

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/GC-0704_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart