KEY181740394 Key Organics / BIONET

1-(5-chloropyridin-2-yl)-4-methyl-4,5-dihydro-1H-pyrazol-3-amine

  • MDL number MFCD11100246
  • Cas Number : 85964-18-7
  • Molecular Weight : 210.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 159 - 161
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97349

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/GC-0752_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart