KEY181740395 Key Organics / BIONET

5,6-dichloropyridine-3-carbonitrile

  • MDL number MFCD11100247
  • Cas Number : 65189-15-3
  • Molecular Weight : 173

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 146 - 148
Iupac Name 5,6-dichloronicotinonitrile
Inchi Code 1S/C6H2Cl2N2/c7-5-1-4(2-9)3-10-6(5)8/h1,3H
Inchi Key JBMLYEZUHCZNKT-UHFFFAOYSA-N

Description

Melting Point 146 - 148
Purity 95%
Storage temp. Store at 4
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97351

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404,P411
MSDS https://www.keyorganics.net/media/catalog/product/sds/GC-0755_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart