KEY181740400 Key Organics / BIONET

methyl 2-methyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-7-carboxylate

  • MDL number MFCD11100233
  • Cas Number : 179950-69-7
  • Molecular Weight : 221.212

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 169 - 171
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97326

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/GC-0724_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart