KEY181740407 Key Organics / BIONET

4-chloro-2-(pyridin-2-yl)-6-(trifluoromethyl)pyrimidine

  • MDL number MFCD11100198
  • Cas Number : 438249-84-4
  • Molecular Weight : 259.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 46 - 47
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70365

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3E-367S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart