KEY181740410 Key Organics / BIONET

5-fluoro-1H-indazol-3-amine

  • MDL number MFCD03426699
  • Cas Number : 61272-72-8
  • Molecular Weight : 151.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 165 - 167
Purity 95
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94516

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/media/catalog/product/sds/AC-5024_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart