KEY181740412 Key Organics / BIONET

tert-butyl 2-amino-5H,6H,7H,8H-pyrido[4,3-d]pyrimidine-6-carboxylate

  • MDL number MFCD08447402
  • Cas Number : 869198-95-8
  • Molecular Weight : 250.302

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170 -172
Purity 95
Storage temp. RT
Physical Form Off-white to Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/95018

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/BC-5021_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart