KEY181740414 Key Organics / BIONET

methyl 3,4-dihydro-2H-1,4-benzoxazine-6-carboxylate

  • MDL number MFCD08544342
  • Cas Number : 758684-29-6
  • Molecular Weight : 193.202

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 69 - 71
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97344

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/GC-0747_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart