KEY181740418 Key Organics / BIONET

(5-cyano-2-methoxyphenyl)boronic acid

  • MDL number MFCD06801691
  • Cas Number : 612833-37-1
  • Molecular Weight : 176.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 211 - 212
Iupac Name 5-cyano-2-methoxyphenylboronic acid
Inchi Code 1S/C8H8BNO3/c1-13-8-3-2-6(5-10)4-7(8)9(11)12/h2-4,11-12H,1H3
Inchi Key LDZCPLGOLLHOON-UHFFFAOYSA-N

Description

Melting Point 211 - 212
Purity 95%
Storage temp. Store under N2
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97319

Safety Information

signalword Warning
pcodes P422, P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/GC-0707_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart