KEY181740419 Key Organics / BIONET

3,5-difluoropyridine-4-carboxylic acid

  • MDL number MFCD04972829
  • Cas Number : 903522-29-2
  • Molecular Weight : 159.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3,5-difluoroisonicotinic acid
Inchi Code 1S/C6H3F2NO2/c7-3-1-9-2-4(8)5(3)6(10)11/h1-2H,(H,10,11)
Inchi Key DRWDDFVJHGAJTN-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97350

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P422,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/GC-0754_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart