KEY182144864 Key Organics / BIONET

methyl 3-oxo-2-propanoylpentanoate

  • MDL number MFCD11553015
  • Cas Number : 158511-22-9
  • Molecular Weight : 186.207

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point OIL
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/3202

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10T-0632_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart