KEY182144865 Key Organics / BIONET

methyl 2-amino-6-methyl-4H,5H,6H,7H-thieno[2,3-c]pyridine-3-carboxylate hydrate

  • MDL number MFCD03701479
  • Cas Number : 449778-52-3
  • Molecular Weight : 244.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105 - 108
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7944

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12F-922_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart