KEY182144883 Key Organics / BIONET

methyl 4-fluoro-2-(trifluoromethyl)benzoate

  • MDL number MFCD06203708
  • Cas Number : 220141-23-1
  • Molecular Weight : 222.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point OIL
Purity 97
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/100917

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/KC-0725_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart