KEY182144896 Key Organics / BIONET

3-{[1,1'-biphenyl]-4-yl}-3-oxopropanenitrile

  • MDL number MFCD02260810
  • Cas Number : 78443-35-3
  • Molecular Weight : 221.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 116 - 118
Iupac Name 3-[1,1'-biphenyl]-4-yl-3-oxopropanenitrile
Inchi Code 1S/C15H11NO/c16-11-10-15(17)14-8-6-13(7-9-14)12-4-2-1-3-5-12/h1-9H,10H2
Inchi Key BZAJCNCPXHAXBZ-UHFFFAOYSA-N

Description

Melting Point 116 - 118
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97302

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/GC-0212_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart