KEY182144904 Key Organics / BIONET

4-(4-chlorophenyl)-3-methyl-1H-pyrazole

  • MDL number MFCD00179386
  • Cas Number : 667400-41-1
  • Molecular Weight : 192.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 155 - 156
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73534

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3W-0884_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart