KEY183157421 Key Organics / BIONET

{4-[3-(trifluoromethyl)pyridin-2-yl]phenyl}methanol

  • MDL number MFCD11501048
  • Cas Number : 1092352-38-9
  • Molecular Weight : 253.224

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 85 - 86
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97364

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/GC-0772_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart