KEY183157434 Key Organics / BIONET

7-bromo-5-methoxy-2-methyl-1,3-benzoxazole

  • MDL number MFCD11553059
  • Cas Number : 1092352-82-3
  • Molecular Weight : 242.072

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105 - 107
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/100929

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/KC-0749_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart