KEY183157445 Key Organics / BIONET

N-benzyl-1H-pyrazolo[3,4-d]pyrimidin-4-amine

  • MDL number MFCD07654603
  • Cas Number : 58360-86-4
  • Molecular Weight : 225.255

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 215 - 217
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/100904

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/KC-0702_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart