KEY183988933 Key Organics / BIONET

2-amino-4-(benzylsulfanyl)-2-methylbutanoic acid

  • MDL number MFCD11553078
  • Cas Number : 19361-15-0
  • Molecular Weight : 239.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108204

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-2978_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart