KEY183988937 Key Organics / BIONET

1-bromo-3-(chloromethyl)-2-fluorobenzene

  • MDL number MFCD09842443
  • Cas Number : 786652-60-6
  • Molecular Weight : 223.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 59 - 61
Iupac Name 1-bromo-3-(chloromethyl)-2-fluorobenzene
Inchi Code 1S/C7H5BrClF/c8-6-3-1-2-5(4-9)7(6)10/h1-3H,4H2
Inchi Key KQLBOTQKXUSZMJ-UHFFFAOYSA-N

Description

Melting Point 59 - 61
Purity 95%
Physical Form Low Melting Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/100935

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/KC-0761_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart