KEY183988942 Key Organics / BIONET

tert-butyl 4-[(4-cyanopyridin-2-yl)oxy]piperidine-1-carboxylate

  • MDL number MFCD09877713
  • Cas Number : 1065484-24-3
  • Molecular Weight : 303.362

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 123 - 124
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/101761

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/LC-0714_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart