KEY183988944 Key Organics / BIONET

3-{2-[(4-chlorophenyl)methoxy]phenyl}-1H-pyrazole

  • MDL number MFCD11553022
  • Cas Number : 1177093-12-7
  • Molecular Weight : 284.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 108 - 110
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/80161

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5R-0224_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart