KEY183988945 Key Organics / BIONET

(E)-2-chloro-N'-hydroxypyridine-4-carboximidamide

  • MDL number MFCD11052368
  • Cas Number : 857653-85-1
  • Molecular Weight : 171.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 162 - 164
Iupac Name 2-chloro-N'-hydroxy-4-pyridinecarboximidamide
Inchi Code 1S/C6H6ClN3O/c7-5-3-4(1-2-9-5)6(8)10-11/h1-3,6H,8H2
Inchi Key GTZXNHAYHRCOHW-UHFFFAOYSA-N

Description

Melting Point 162 - 164
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/100931

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/KC-0753_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart