KEY183988951 Key Organics / BIONET

[(2E)-3-ethoxy-2-(methoxyimino)-3-oxopropyl]triphenylphosphanium bromide

  • MDL number MFCD11553020
  • Cas Number : 108411-45-6
  • Molecular Weight : 486.346

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76208

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4P-305S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart