KEY183988952 Key Organics / BIONET

ethyl 6-bromoimidazo[1,2-a]pyridine-2-carboxylate hydrobromide

  • MDL number MFCD09991793
  • Cas Number : 67625-37-0
  • Molecular Weight : 350.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/96365

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/EB-0851_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart