KEY183988956 Key Organics / BIONET

2-(furan-2-yl)-5-hydroxy-1-benzofuran-3-carbaldehyde

  • MDL number MFCD11553023
  • Cas Number : 1092352-20-9
  • Molecular Weight : 228.203

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 213 - 215
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84469

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6Y-0818_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart