KEY183988970 Key Organics / BIONET

(2E)-3-ethoxy-2-methanesulfonylprop-2-enenitrile

  • MDL number MFCD11553018
  • Cas Number : 104007-26-3
  • Molecular Weight : 175.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73081

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3T-0083_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart