KEY183988972 Key Organics / BIONET

4-(benzyloxy)-3-bromobenzaldehyde

  • MDL number MFCD02629356
  • Cas Number : 69455-12-5
  • Molecular Weight : 291.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 96 - 99
Iupac Name 4-(benzyloxy)-3-bromobenzaldehyde
Inchi Code 1S/C14H11BrO2/c15-13-8-12(9-16)6-7-14(13)17-10-11-4-2-1-3-5-11/h1-9H,10H2
Inchi Key LMAYCCCBNRUZPC-UHFFFAOYSA-N

Description

Melting Point 96 - 99
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/101768

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/LC-0725_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart