KEY183988973 Key Organics / BIONET

4-bromo-1-isoquinolinamine

  • MDL number MFCD05155261
  • Cas Number : 55270-27-4
  • Molecular Weight : 223.073

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108211

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-2988_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart