KEY183988975 Key Organics / BIONET

2-[2-(benzyloxy)-2-oxoethyl]acrylic acid

  • MDL number MFCD11553080
  • Cas Number : 48162-88-5
  • Molecular Weight : 220.224

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108209

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-2986_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart