KEY183988976 Key Organics / BIONET

N'-hydroxy-5,6-dimethoxy-1-benzothiophene-2-carboximidamide hydrochloride

  • MDL number MFCD11553077
  • Cas Number : 101041-95-6
  • Molecular Weight : 288.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 85
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/108202

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-2975_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart