KEY183988985 Key Organics / BIONET

N1-methyl-4-nitrobenzene-1,2-diamine

  • MDL number MFCD00156607
  • Cas Number : 41939-61-1
  • Molecular Weight : 167.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 174 - 176
Iupac Name N~1~-methyl-4-nitro-1,2-benzenediamine
Inchi Code 1S/C7H9N3O2/c1-9-7-3-2-5(10(11)12)4-6(7)8/h2-4,9H,8H2,1H3
Inchi Key MNIKERWISBANET-UHFFFAOYSA-N

Description

Melting Point 174 - 176
Purity 97%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/101763

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/LC-0716_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart