KEY183988987 Key Organics / BIONET

methyl 6-nitro-2-oxo-2H-chromene-3-carboxylate

  • MDL number MFCD00488482
  • Cas Number : 91059-67-5
  • Molecular Weight : 249.178

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 212 - 214
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94330

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9Y-0806_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart