KEY184991887 Key Organics / BIONET

2-cyclopropyl-5-hydroxy-1-benzofuran-3-carbaldehyde

  • MDL number MFCD11840949
  • Cas Number : 1135282-80-2
  • Molecular Weight : 202.209

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 194 - 196
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84458

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6Y-0800_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart