KEY184991889 Key Organics / BIONET

3-chloro-2-iodo-5-nitropyridine

  • MDL number MFCD11840993
  • Cas Number : 488713-29-7
  • Molecular Weight : 284.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 74 - 76
Iupac Name 3-chloro-2-iodo-5-nitropyridine
Inchi Code 1S/C5H2ClIN2O2/c6-4-1-3(9(10)11)2-8-5(4)7/h1-2H
Inchi Key FCXUHIMBZWWQMB-UHFFFAOYSA-N

Description

Melting Point 74 - 76
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97307

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/GC-0608_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart