KEY184991893 Key Organics / BIONET

2-amino-5-chloropyridine-3-carbaldehyde

  • MDL number MFCD10696877
  • Cas Number : 54856-61-0
  • Molecular Weight : 156.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 169 - 171
Purity 97
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97328

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/GC-0727_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart