KEY184991910 Key Organics / BIONET

[5-amino-2-(morpholin-4-yl)phenyl]methanol

  • MDL number MFCD11841019
  • Cas Number : 759456-60-5
  • Molecular Weight : 208.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 133 - 135
Iupac Name [5-amino-2-(4-morpholinyl)phenyl]methanol
Inchi Code 1S/C11H16N2O2/c12-10-1-2-11(9(7-10)8-14)13-3-5-15-6-4-13/h1-2,7,14H,3-6,8,12H2
Inchi Key VUCXPUVUVWCVDL-UHFFFAOYSA-N

Description

Melting Point 133 - 135
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/101773

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/LC-0736_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart