KEY184991913 Key Organics / BIONET

4-iodo-1H-indazol-3-amine

  • MDL number MFCD03426697
  • Cas Number : 599191-73-8
  • Molecular Weight : 259.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 154 - 155 C
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97306

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/GC-0607_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart