KEY184991921 Key Organics / BIONET

5-amino-1-[2-nitro-4-(trifluoromethyl)phenyl]-1H-pyrazole-4-carbonitrile

  • MDL number MFCD00205968
  • Cas Number : 74990-43-5
  • Molecular Weight : 297.197

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 231 - 233
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/100903

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/KC-0701_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart