KEY184991923 Key Organics / BIONET

tert-butyl 4-(aminomethyl)-4-hydroxypiperidine-1-carboxylate

  • MDL number MFCD11579803
  • Cas Number : 392331-66-7
  • Molecular Weight : 230.308

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 148 - 150
Purity 95
Storage temp. Store under N2
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/97332

Safety Information

signalword Warning
pcodes P422, P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/GC-0732_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart