KEY184991961 Key Organics / BIONET

6-(4-propoxyphenoxy)pyridine-3-carbonitrile

  • MDL number MFCD10693376
  • Cas Number : 1135282-78-8
  • Molecular Weight : 254.289

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73844

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3Y-5030_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart