KEY184991987 Key Organics / BIONET

(5-fluoro-1-benzothiophen-2-yl)methanol

  • MDL number MFCD11841046
  • Cas Number : 13771-73-8
  • Molecular Weight : 182.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 91 - 93
Iupac Name (5-fluoro-1-benzothien-2-yl)methanol
Inchi Code 1S/C9H7FOS/c10-7-1-2-9-6(3-7)4-8(5-11)12-9/h1-4,11H,5H2
Inchi Key MLMIMHHVDSBESY-UHFFFAOYSA-N

Description

Melting Point 91 - 93
Purity 95%
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/106710
Availability details for
Quantity
  • Availability
  • Add to Cart