KEY184992014 Key Organics / BIONET

3-{[(tert-butoxy)carbonyl]amino}pyridine-4-carboxylic acid

  • MDL number MFCD03427723
  • Cas Number : 179024-65-8
  • Molecular Weight : 238.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 280 C (dec)
Iupac Name 3-[(tert-butoxycarbonyl)amino]isonicotinic acid
Inchi Code 1S/C11H14N2O4/c1-11(2,3)17-10(16)13-8-6-12-5-4-7(8)9(14)15/h4-6H,1-3H3,(H,13,16)(H,14,15)
Inchi Key ZZXUPXUHQHTUNH-UHFFFAOYSA-N

Description

Melting Point 280 C (dec)
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/106702

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/NC-0713_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart