KEY184992017 Key Organics / BIONET

(3E)-4-(4-fluorophenyl)but-3-en-2-one

  • MDL number MFCD00205144
  • Cas Number : 1611-38-7
  • Molecular Weight : 164.179

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 119 C @ 2.5mm Hg
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63276

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1D-016_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart