KEY184992029 Key Organics / BIONET

6-[2-(4-methyl-1,3-thiazol-5-yl)ethoxy]pyridine-3-carbonitrile

  • MDL number MFCD09940701
  • Cas Number : 1016875-35-6
  • Molecular Weight : 245.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73851

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3Y-5070_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart